Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. F078925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F078925G 25 g
F07895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F078925G Supplier TCI America Supplier No. F078925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2369-11-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00034065
InChI Key PEDMFCHWOVJDNW-UHFFFAOYSA-N
Synonym 2-nitro-5-fluoroaniline, 5-fluoro-2-nitrophenylamine, 2-amino-4-fluoronitrobenzene, 3-fluoro-6-nitroaniline, 5-fluoro-2-nitrobenzenamine, benzenamine, 5-fluoro-2-nitro, pubchem4476, acmc-1cegf, 5-fluoro-2-nitro-aniline, intermediates-zcf02632
PubChem CID 75399
IUPAC Name 5-fluoro-2-nitroaniline
SMILES C1=CC(=C(C=C1F)N)[N+](=O)[O-]

Specifications

Melting Point 98°C
Quantity 25 g
Formula Weight 156.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-2-nitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.