Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-nitrobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. F049725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F049725G 25 g
F04975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F049725G Supplier TCI America Supplier No. F049725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 393-09-9
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00039741
InChI Key WMQOSURXFLBTPC-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
PubChem CID 223094
IUPAC Name 4-fluoro-1-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 g
Formula Weight 209.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-2-nitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.