Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-8-methoxyquinoline 98.0+%, TCI America™
SDP

Catalog No. F09091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09091G 1 g
F09095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09091G Supplier TCI America Supplier No. F09091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 439-88-3
Molecular Formula C10H8FNO
Molecular Weight (g/mol) 177.178
MDL Number MFCD18452297
InChI Key XNOHPRHYFXAKPA-UHFFFAOYSA-N
PubChem CID 89345024
IUPAC Name 5-fluoro-8-methoxyquinoline
SMILES COC1=C2C(=C(C=C1)F)C=CC=N2

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 147°C
Quantity 1 g
Formula Weight 177.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-8-methoxyquinoline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.