Learn More
Cayman Chemical JNK InhIbItr IX 25mg
SDP
Supplier:  Cayman Chemical 1562425 MG
Encompass_Preferred
An analog of cyclopamine that inhibits the Hh signaling-dependent proliferation of NIH/3T3 cells at 8 M and dose dependently inhibits platelet aggregation in rabbits ex vivo induces serotonin release and inhibits its re-uptake in the CNS
Catalog No. 50-297-0606
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.