Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorobenzoxazole, 97%, Thermo Scientific™

Catalog No. AC455610010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455610010 1 g
AC455612500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455610010 Supplier Thermo Scientific Chemicals Supplier No. 455610010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 221347-71-3
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.11
InChI Key ZRMPAEOUOPNNPZ-UHFFFAOYSA-N
Synonym 5-fluorobenzoxazole, 5-fluorobenzo d oxazole, benzoxazole, 5-fluoro, benzoxazole,5-fluoro, pubchem21888, 5-fluoranyl-1,3-benzoxazole, benzoxazole, 5-fluoro-9ci
PubChem CID 19937167
IUPAC Name 5-fluoro-1,3-benzoxazole
SMILES C1=CC2=C(C=C1F)N=CO2

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 137.11
Percent Purity 97%
Chemical Name or Material 5-Fluorobenzoxazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.