Learn More
Medchemexpress LLC 5-fluorocytidine | 2341-22-2 | MFCD00210953 | 97.0% | 261.21 g/mol | C9H12FN3O5 | 1 G

Supplier: Medchemexpress LLC HYW0397221G
5-fluorocytidine is a fluorinated cytidine nucleoside that inhibits maturation of the 45S ribosomal RNA precursor and is provided as a characterized research reagent with storage and safety guidance from product datasheets and SDS.
- Fluorinated cytidine nucleoside.
- Inhibits maturation of the 45S ribosomal RNA precursor.
- Molecular formula C9H12FN3O5.
- Molecular weight 261.21 g/mol.
- Storage: 4°C under nitrogen; in solvent: -80°C for 6 months, -20°C for 1 month.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.