Learn More
Cayman Chemical 5-FluorocytosIn 5g
SDP
Supplier:  Cayman Chemical 116355 G
Encompass_Preferred
A reversble 5-LO nhbtor nhbts 5-LO actvty n RBL-1 cell supernatant (C50 0 5 UM) nhbts LTB4 producton nduced by A23187 n purfed human PMNLs (C50 0 6 UM) prevents antgen challenge-nduced ncreases n specfc lung resstance n a sheep model of asthma at 4 to 8 hours post-challenge at 10 mg/kg p o 2 hours pre-challenge
Catalog No. 50-295-4155
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.