Learn More
AdipoGen 5-Fluorocytosine
SDP
Supplier:  AdipoGen CDXF0077G005
Encompass_Preferred
Chemical. CAS 2022-85-7. Formula C4H4FN3O. MW 129.09. Synthetic Fluorinated pyrimidine analog. Antimycotic prodrug that is converted by cytosine deaminase to 5-fluorouracil a widely used cytotoxic drug and further metabolized to fluorinated ribo- and deoxyribonucleotides. Inhibits DNA and RNA synthesis and interfers with ribosomal protein synthesis. Displays antifungal and antitumor activity. Lately with the development of gene therapy, it has been introduced as a prodrug in combination with the cytosine deaminase suicide gene as antitumor compound, significantly improving survival and reducing tumor size in selected tumors.
Catalog No. 50-256-0898
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.