Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorodeoxyuridine, ≥98%, MP Biomedicals™

Catalog No. ICN10555180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
0.25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10555180 0.1 g
ICN10555125 25 mg
ICN10555183 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10555180 Supplier MP Biomedicals, Inc Supplier No. 0210555180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Color White
Assay Percent Range ≥98%
Quantity 0.1 g
Formula Weight 246.19
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorodeoxyuridine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.