Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. F07381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07381G 1 g
F07385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07381G Supplier TCI America Supplier No. F07381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 402-66-4
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.10
MDL Number MFCD01318537
InChI Key BXZSBDDOYIWMGC-UHFFFAOYSA-N
Synonym 5-fluoronicotinic acid, 5-fluoro-3-pyridinecarboxylic acid, 5-fluoro-nicotinic acid, 3-pyridinecarboxylic acid, 5-fluoro, nicotinic acid, 5-fluoro, 3-fluoro-5-carboxypyridine, 3-fluoro-5-pyridinecarboxylic acid, 5-fluoronicotinicacid, pubchem10942
PubChem CID 120232
IUPAC Name 5-fluoropyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(F)=C1

Specifications

Melting Point 199°C
Color White-Yellow
Quantity 1 g
Formula Weight 141.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoronicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.