Learn More
Abcam 5-Fluoroorotic Acid Hydrate, >99% Purity, Solid, 1 g
Description
- Purity: >99%
- Molecular Weight: 174.1 g/mol
- Storage Temperature: Store at -20°C
- Shipping Temperature: Ambient - Can Ship with Ice
- Form: Solid
- Applications: Selective agent in yeast molecular genetics, useful in the selection of orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae
- Composition: 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid; hydrate
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Orotidine-5¹-phosphate Decarboxylase |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid, hydrate |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate |
| Quantity | 1 g |
| For Use With (Application) | Yeast molecular genetics, selection of orotidine-5'-phosphate decarboxylase mutants |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.