Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 5-Fluoroorotic Acid Hydrate, Solid, >99% Purity, 5 g

Catalog No. 502704802
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4802 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4802 Supplier Abcam Supplier No. AB1462455G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity chemical reagent for the selection of orotidine-5'-phosphate decarboxylase mutants in yeast molecular genetics.

  • Molecular weight: 174.1 g/mol
  • Purity: >99%
  • Form: Solid
  • CAS Number: 207291-81-4
  • Description: Selective agent in yeast molecular genetics, useful in the selection of orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
  • Storage: Store at -20°C.
  • Constituents: 100% 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Content And Storage Store at -20°C.
Form Solid
Target Molecule Orotidine-5'-phosphate Decarboxylase
Quantity 5 g
For Use With (Application) Yeast molecular genetics, selection of orotidine-5'-phosphate decarboxylase mutants
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.