Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroorotic Acid Monohydrate 95.0+%, TCI America™
SDP

Catalog No. F0382100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0382100MG 100 mg
F03821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0382100MG Supplier TCI America Supplier No. F0382100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 220141-70-8
Molecular Formula C5H5FN2O5
Molecular Weight (g/mol) 192.10
MDL Number MFCD00150658
InChI Key LODRRYMGPWQCTR-UHFFFAOYSA-N
Synonym 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, 5-fluoroorotic acid monohydrate, 5-fluoroorotic acid hydrate, 5-fluoroorotic acid,monohydrate, 5-fluoro-1,2,3,6-tetrahydro-2,6-dioxopyrimidine-4-carboxylic acid hydrate, 5-fluoroortic acid monohydrate, 5-fluorouracil-4-carboxylic acid hydrate, 5-fluoroorotic acid n-hydrate, 5-fluororotic acid monohydrate, 5-fluoro-orotic acid monohydrate
PubChem CID 16212749
IUPAC Name 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate
SMILES O.OC(=O)C1=C(F)C(=O)NC(=O)N1

Specifications

Quantity 100 mg
Formula Weight 174.09
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroorotic Acid Monohydrate
RTECSNumber UV7800000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.