Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluorosalicylaldehyde 98.0+%, TCI America™
SDP

Catalog No. F057425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F057425G 25 g
F05745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F057425G Supplier TCI America Supplier No. F057425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 347-54-6
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD01090997
InChI Key FDUBQNUDZOGOFE-UHFFFAOYSA-N
Synonym 5-fluorosalicylaldehyde, 5-fluoro-2-hydroxy-benzaldehyde, benzaldehyde, 5-fluoro-2-hydroxy, 5-fluoro-2-hydroxy benzaldehyde, pubchem1444, 5-fluoro salicylaldehyde, 5-?fluorosalicylaldehyde, acmc-209ia4, ksc492m4t, pharmabridge p-2996
PubChem CID 2737328
IUPAC Name 5-fluoro-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)O

Specifications

Melting Point 85°C
Color White-Yellow
Boiling Point 56°C
Quantity 25 g
Formula Weight 140.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluorosalicylaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.