Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 5-Fluorouracil
GREENER_CHOICESDP

Catalog No. 325750
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-575-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 32-575-0 Supplier Tocris Bioscience™ Supplier No. 3257/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thymidylate synthetase inhibitor

Anticancer agent. Metabolized to form fluorodeoxyuridine monophosphate (FdUMP), fluorodeoxyuridine triphosphate (FdUTP) and fluorouridine (FUTP). FdUMP inhibits thymidylate synthase, causing a reduction in dTMP synthesis. FUTP and FdUTP are misincorporated into RNA and DNA respectively.

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2

Specifications

Quantity 50 mg
Formula Weight 130.08
Purity >99%
Molecular Weight (g/mol) 130.08
Chemical Name or Material 5-Fluorouracil
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.