Learn More
Apexbio Technology LLC 5-Formyl-dCTP(Synonyms: 5-Formyl-2'-deoxycytidine-5'-triphosphate, 5f-dCTP, fdCTP), 50ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B81170.05
Encompass_Preferred
5-Formyl-dCTP is a modified cytosine-based deoxynucleoside triphosphate derivative capable of incorporation into DNA by DNA polymerase activity resulting in the formation of 5-formylcytosine (5fC) modified bases Presence of 5fC marks an intermediate step within active DNA demethylation pathways impacting epigenetic regulatory processes and gene expression It serves as a reagent in DNA modification and epigenomic studies facilitating investigations into the functional roles of 5fC in replication fidelity base excision repair mechanisms transcriptional regulation and chromatin remodeling dynamics
Catalog No. 50-000-60898
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.