Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formylsalicylic Acid 98.0+%, TCI America™
SDP

Catalog No. F040025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F040025G 25 g
F04005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F040025G Supplier TCI America Supplier No. F040025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-76-2
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00006945
InChI Key UTCFOFWMEPQCSR-UHFFFAOYSA-N
Synonym 5-formylsalicylic acid, 2-hydroxy-5-formylbenzoic acid, benzoic acid, 5-formyl-2-hydroxy, 5-formylsalicylicacid, 5-formylsalicylic acide, 3-carboxy-4-hydroxybenzaldehyde, 5-formyl-2-hydroxy-benzoic acid, pubchem10569, acmc-209mva, 5-formyl salicylic acid
PubChem CID 69226
IUPAC Name 5-formyl-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(C=O)=CC=C1O

Specifications

Melting Point 250°C
Color Yellow
Quantity 25 g
Formula Weight 166.13
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Formylsalicylic Acid
EINECSNumber (3)-1669
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.