Learn More
Labconco™ 5 ft. Cell Logic™ Class II, Type A2 Biosafety Cabinet, US Model
Shop All Labconco Corporation Products

Description
Performance Features:
- Safe Class II airflows (approx. 70% recirculating, 30% exhausted) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe cabinet airflows – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display.
- Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
- Stand-Still™ Isolation Platform is required (sold separately).
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
- Water circulator is required (not included).
- Telescoping base stand with shelf and leveling feet.
Specifications
Specifications
| Description | 5 ft. Cell Logic Class II, Type A2 Biosafety Cabinet |
| Certifications/Compliance | ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 |
| Chamber Material | Stainless Steel (304) |
| Width (Metric) Interior | 153.7 cm |
| Product Type | Class II, Type A2 Bio Safety Cabinet |
| Model | Cell Logic |
| Noise Level | 63 dBA |
| Outlets | 2 Duplex |
| Plug Type | North America (115 V, 20 A) |
| Power Consumption | 240 W |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.