Learn More
Lime Media 5 Gallon Hand Sanitizer Floor Stand with Stainless Steel Touchless Dispenser
Description
- Stainless Steel Touchless Bulk-fill Dispenser
- Aluminum enclosed one-piece stand
- 5 gallon interior capacity
- Available in black and white powder-coated finishes
- Indoor/outdoor compatible
- Pre-drilled base hole for mounting
Specifications
Specifications
| Product Type | Sanitizer Dispenser Stand |
| Color | White |
| Sterility | Non-sterile |