Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexen-1-ol 97.0+%, TCI America™
SDP

Catalog No. H0653250ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
25 mL
250 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0653250ML 250 mL
H065310ML 10 mL
H065325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0653250ML Supplier TCI America Supplier No. H0653250ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 821-41-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00002981
InChI Key UIZVMOZAXAMASY-UHFFFAOYSA-N
Synonym 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
PubChem CID 69963
IUPAC Name hex-5-en-1-ol
SMILES C=CCCCCO

Specifications

Color Colorless
Boiling Point 157°C
Quantity 250 mL
UN Number 1987
Formula Weight 100.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.