Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexenenitrile 95.0+%, TCI America™
SDP

Catalog No. H083625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H083625ML 25 mL
H08365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H083625ML Supplier TCI America Supplier No. H083625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5048-19-1
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.145
MDL Number MFCD00067056
InChI Key UNAQSRLBVVDYGP-UHFFFAOYSA-N
Synonym 5-hexenenitrile, 5-cyano-1-pentene, 4-pentenyl cyanide, hex-1-ene-6-nitrile, acmc-209kmu
PubChem CID 123222
IUPAC Name hex-5-enenitrile
SMILES C=CCCCC#N

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 mL
UN Number 1992
Formula Weight 95.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexenenitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.