Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexenyl acetate, 97%

Catalog No. p-7035719
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5344506 5 g
AAH5344514 25 g
AAH5344522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5344506 Supplier Thermo Scientific Chemicals Supplier No. H5344506
Only null left
Add to Cart

CAS: 5048-26-0 | C8H14O2 | 142.20 g/mol

(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4E,7Z)-4,7-Tridecadienyl acetate,a component of the sex pheromone was synthesized from 5-hexenyl acetate. CM of 5-decene with 5-hexenyl acetate is used to produce the insect pheromone. Used in olefin cross metathesis.

Solubility
Insoluble in water, soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5048-26-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00060856
InChI Key MPLWNENKBSBMFN-UHFFFAOYSA-N
Synonym 5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
PubChem CID 537536
IUPAC Name hex-5-enyl acetate
SMILES CC(=O)OCCCCC=C

Specifications

Density 0.883
Boiling Point 173°C to 174°C
Flash Point 60°C (142°F)
Refractive Index 1.423
Linear Formula CH3CO2(CH2)4CH=CH2
Quantity 5 g
UN Number UN3272
Solubility Information Insoluble in water,soluble in alcohol.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 5-Hexenyl acetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.