Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexenyl Acetate 98.0+%, TCI America™
SDP

Catalog No. A1338500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1338500ML 500 mL
A133825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1338500ML Supplier TCI America Supplier No. A1338500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5048-26-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00060856
InChI Key MPLWNENKBSBMFN-UHFFFAOYSA-N
Synonym 5-hexenyl acetate, hex-5-en-1-yl acetate, acetic acid 5-hexenyl ester, 5-hexen-1-ol, acetate, 5-hexene-1-ol, acetate, acetic acid 5-hexen-1-yl ester, 6-acetoxy-1-hexene, 6-acetoxyhex-1-ene, 5-hexenyl acetate #, acmc-1aw0j
PubChem CID 537536
IUPAC Name hex-5-en-1-yl acetate
SMILES CC(=O)OCCCCC=C

Specifications

Color Colorless
Boiling Point 172°C
Quantity 500 mL
UN Number 3272
Formula Weight 142.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexenyl Acetate
EINECSNumber (2)-0760&(2)-2533
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.