Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexyn-1-ol 95.0+%, TCI America™
SDP

Catalog No. H068725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H068725ML 25 mL
H06875ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H068725ML Supplier TCI America Supplier No. H068725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 928-90-5
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00002980
InChI Key GOQJMMHTSOQIEI-UHFFFAOYSA-N
Synonym 5-hexyn-1-ol, 1-hydroxy-5-hexyne, 5-hexyne-1-ol, 5-hextyn-1-ol, 5-hexynol, 5-hexynyl alcohol, hex-5-ynol, 5-hexyn-l-ol, 6-hydroxy-1-hexyne, pubchem13078
PubChem CID 70234
IUPAC Name hex-5-yn-1-ol
SMILES OCCCCC#C

Specifications

Color Yellow
Boiling Point 75°C
Quantity 25 mL
Formula Weight 98.15
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Hexyn-1-ol
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.