Learn More
Medchemexpress LLC 5-HMSiR-Hochest | 2996108-30-4 | 98.7% | 908.17 g/mol | C54H57N9O3Si | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD222210MG
Encompass_Preferred
5-HMSiR-Hoechst is a far-red fluorescent DNA probe that links a DNA minor-groove binder to a spontaneously blinking hydroxymethyl silicon-rhodamine (HMSiR) fluorophore. It is designed for live-cell nuclear staining and imaging applications that benefit from far-red excitation and blinking behavior.
  • Provides far-red excitation/emission (640 nm/675 nm) suitable for multicolor imaging.
  • Compatible with live-cell staining and reduced phototoxicity.
  • Spontaneously blinking fluorophore supports super-resolution and single-molecule imaging.
  • High chemical purity for reliable imaging performance.
  • Offered in small research sizes for assay development and optimization.
Catalog No. 50-002-97414