Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HTR3 Polyclonal Antibody, Bioss
SDP

Catalog No. 502280553
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0553 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0553 Supplier Bioss Supplier No. BS34076R
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

5-HTR3 Polyclonal antibody specifically detects 5-HTR3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen 5-HTR3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Htr3a
Gene Accession No. P23979, P35563, P46098
Gene Alias 5HT3; 5-HT3; 5-HT-3; 5-HT3 receptor; 5HT3 serotonin receptor; 5-HT3A; 5-HT3-A; 5HT3R; 5-HT3R; 5-hydroxytryptamine (serotonin) receptor 3A; 5-hydroxytryptamine (serotonin) receptor 3A, ionotropic; 5-hydroxytryptamine receptor 3; 5-hydroxytryptamine receptor 3A; Htr3; Htr3a; serotonin receptor; Serotonin receptor 3A; serotonin-gated ion channel receptor; truncated receptor
Host Species Rabbit
Immunogen Recombinant human 5-HTR3
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15561, 3359, 79246
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.