Learn More
Medchemexpress LLC 5α-hydroxycostic acid | 132185-83-2 | MFCD20260447 | 95.0% | 250.33 g/mol | C15H22O3 | 10 MG

Supplier: Medchemexpress LLC HYN266610MG
5α-Hydroxycostic acid is a eudesmane-type sesquiterpene research reagent reported to inhibit angiogenesis through modulation of VEGF/VEGFR2 and Ang2/Tie2 signaling pathways. It is provided as a solid for in vitro research and preclinical studies.
- Natural eudesmane-type sesquiterpene with antiangiogenic activity.
- Inhibits angiogenesis and suppresses cancer cell migration via VEGF/VEGFR2 and Ang2/Tie2 pathways.
- Supplied as a solid suitable for in vitro research applications.
- Purity HPLC 95.0%.
- Molecular formula C15H22O3, molecular weight 250.33 g/mol.
- Recommended storage: protect from light; store at -20°C (in solvent: -80°C for 6 months, -20°C for 1 month).
- Typical package size: 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.