Learn More
Sigma Organic Chemistry 5-Hydroxymethyl-2-fural | 1G | 67-47-0 | MFCD00003234
Supplier: Sigma Organic Chemistry H408071G

5-Hydroxymethyl-2-furaldehyde, 1G
About This Item:
Synonym(s): 5-(Hydroxymethyl)furfural; 5-Hydroxymethyl-2-furancarboxaldehyde; HMF
Linear Formula: C6H6O3
Molecular Weight: 126.11
CAS Number: 67-47-0
MDL Number: MFCD00003234
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 114-116 C/1 mmHg (lit.)
Melting Point: 28-34 C (lit.)
Density: 1.243 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.562 (lit.)
Storage: 2-8C
EINECS Number: 200-654-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.