Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxypyridine-2-carboxylic acid, 98+%

Catalog No. AAH6426506
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6426506 5 g
AAH6426503 1 g
AAH6426514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6426506 Supplier Thermo Scientific Chemicals Supplier No. H6426506
Add to Cart

CAS: 15069-92-8 | C6H5NO3 | 139.11 g/mol

Useful synthetic intermediate. Also intermediate for pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15069-92-8
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00661309
InChI Key CGRRUFNHHQCLDZ-UHFFFAOYSA-N
Synonym 5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
PubChem CID 268740
IUPAC Name 5-hydroxypyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1O)C(=O)O

Specifications

Melting Point 260°C to 264°C
Quantity 5 g
Beilstein 116009
Solubility Information Soluble in acetone,dimethyl sulfoxide ,ethanol and methanol.
Formula Weight 139.11
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 5-Hydroxypyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.