Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Isocyanato-1-methyl-1H-indole, 97%, Thermo Scientific™

Catalog No. CC41406CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC41406CB 250 mg Amber Glass Bottle
CC41406DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC41406CB Supplier Thermo Scientific Chemicals Supplier No. CC41406CB
Add to Cart

Chemical Identifiers

CAS 884507-16-8
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD09702358
InChI Key RDQRMCJCROUHOF-UHFFFAOYSA-N
PubChem CID 24229489
SMILES CN1C=CC2=CC(=CC=C12)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-isocyanato-1-methyl-1h-indole, 1h-indole, 5-isocyanato-1-methyl, 1-methylindol-5-isocyanate, 1h-indole,5-isocyanato-1-methyl, 1-methyl-1h-indole-5-yl isocyanate
IUPAC Name 5-isocyanato-1-methylindole
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 5-Isocyanato-1-methyl-1H-indole
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.