Learn More
United States Biological Corporation Glycine Ultrapure, 56-40-6, MFCD00008131, 5kg
Supplier:  United States Biological Corporation G81655KG
Encompass_Preferred
Molecular Formula C2H5NO2, Purity ≥99%, Molecular Weight 75.07, Solubility: Complete solubility in water; slight solubility in alcohol and ether. Synonyms: Aminoacetic acid; Glycocol; Aminoethanoic acid. Glycine is used in Tris-Glycine electrophoresis buffer formulations. This is a highly purified grade suitable for use in peptide synthesis. Special grade of Glycine used specifically for cell culture and Molecular Biology applications.
Catalog No. NC9726283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.