Learn More
Thermo Scientific™ 5 L DynaDrive™ Bioreactor Probe Kit Puck Version
Catalog No. F1102751001 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
5 L
Catalog No. Capacity (Metric)
F1102751001 5 L
1 options
Catalog No. F1102751001 Supplier Thermo Scientific™ Supplier No. F1102751001
Add to Cart
Add to Cart
Request a Quote
DynaDrive bioreactor puck sensor probe kit. Compatible with HyPerforma G3Lab controllers and 5 L DynaDrive bioreactor stands and containers using sensor puck technology. Enables pH and DO monitoring for bioprocess applications.
The Thermo Scientific™ DynaDrive™ 5 L BPC Sensor Puck Kit, F110-2751-001, enables rapid pH and dissolved oxygen (DO) measurement when using sensor puck–configured DynaDrive™ 5 L BioProcess Container (BPC). Designed for compatibility with Thermo Scientific™ HyPerforma™ G3Lab™ Bioprocess Controllers, this kit supports rapid setup, reduced handling, and reliable process monitoring during upstream process development.
The sensor puck kit integrates with the DynaDrive™ 5 L Single-Use Bioreactor ecosystem.
Key features and benefits
  • pH and DO measurement
  • Rapid setup
  • Designed specifically for use with sensor puck–enabled DynaDrive™ 5 L BPC
  • Compatible for use with HyPerforma™ G3Lab™ Bioprocess Controllers to support bioprocess control strategies
Kit includes
  • Puck Sensor Reader (Electro-Optical Module)
  • Cables for Puck Sensor Reader to connect to G3Lab controller
Typical applications
  • Upstream process development and optimization
  • Media and feed screening
  • Scale-down modeling for fed-batch or perfusion processes
Designed for the DynaDrive™ ecosystem
The F110-2751-001 sensor puck kit is part of the Thermo Scientific™ DynaDrive™ Single-Use Bioreactor platform, supporting efficient, scalable, and standardized upstream workflows from early process development through technology transfer.
Specifications
Capacity (Metric) 5 L
Material Stainless Steel (304)
Type Probe Kit