Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5 Lipoxygenase Mouse anti-Human, Porcine, Clone: 3B1B7, Proteintech
SDP

Catalog No. 501736758 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6758 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6758 Supplier Proteintech Group Inc Supplier No. 663261IG150UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT) (Funk, 2001). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma (Drazen et al., 1994), fibrosis (Wilborn et al., 1996) and atherosclerosis (Dwyer et al., 2004). Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO (Luo et al., 2004). Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation. The phospho-specific antibody to Ser523 on 5-LO is thus likely to provide a valuable toll for studies of the role of 5-LO regulation in diseases such as asthma, fibrosis and atherosclerosis.

Specifications

Antigen 5 Lipoxygenase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3B1B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ALOX5
Gene Accession No. P09917
Gene Alias 5 lipoxygenase, 5 LO, 5 LOX, 5LPG, ALOX5, arachidonate 5 lipoxygenase, LOG5
Gene Symbols ALOX5
Host Species Mouse
Immunogen 5 Lipoxygenase Fusion Protein Ag18168
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100156205, 240
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.