Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-mCpA Monoclonal Antibody (2C8H8A6), Active Motif™
SDP

Catalog No. 501993684
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3684 10 μg
50-199-3683 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3684 Supplier Active Motif Supplier No. 61784
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methylation of CpA (mCpA) generates a DNA pyrimidine with a methyl group in the 5-carbon position. DNA methylation is usually catalyzed by DNA methyltransferases generally at CpG dinucleotide positions, though non-CpG methylation has also been reported at CpA and CpT sites. DNA methylation is known to repress gene expression.

Specifications

Antigen 5-mCpA
Applications Dot Blot
Classification Monoclonal
Clone 2C8H8A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Host Species Mouse
Immunogen This antibody was raised against 5-methyl-cytosine-adenosine di-nucleoside conjugated to KLH and recognized 5-mCpA.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Multi-species, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.