Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-1-indanone, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
50g
5111-70-6
C10H10O2
162.19
MFCD00003789
QOPRWBRNMPANKN-UHFFFAOYSA-N
5-methoxy-1-indanone,5-methoxyindan-1-one,5-methoxy-2,3-dihydro-1h-inden-1-one,5-methoxyindanone,2,3-dihydro-5-methoxyinden-1-one,1h-inden-1-one, 2,3-dihydro-5-methoxy,5-methoxy indanone,5-methoxyhydrindone,pubchem2031,1-oxo-5-methoxyindane
78787
5-methoxy-2,3-dihydroinden-1-one
COC1=CC2=C(C=C1)C(=O)CC2
5111-70-6
C10H10O2
162.19
MFCD00003789
QOPRWBRNMPANKN-UHFFFAOYSA-N
5-methoxy-1-indanone,5-methoxyindan-1-one,5-methoxy-2,3-dihydro-1h-inden-1-one,5-methoxyindanone,2,3-dihydro-5-methoxyinden-1-one,1h-inden-1-one, 2,3-dihydro-5-methoxy,5-methoxy indanone,5-methoxyhydrindone,pubchem2031,1-oxo-5-methoxyindane
78787
5-methoxy-2,3-dihydroinden-1-one
COC1=CC2=C(C=C1)C(=O)CC2
Melting Point 107.0°C to 111.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown-Green
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Beilstein 08, II, 152
Formula Weight 162.19
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Methoxy-1-indanone
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-838-6