Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-methylbenzothiazole 98.0+%, TCI America™
SDP

Catalog No. M12815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12815G 5 g
M128125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12815G Supplier TCI America Supplier No. M12815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2941-69-7
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.24
MDL Number MFCD00082664
InChI Key SAQMNBWVOKYKPZ-UHFFFAOYSA-N
PubChem CID 76252
IUPAC Name 5-methoxy-2-methyl-1,3-benzothiazole
SMILES COC1=CC=C2SC(C)=NC2=C1

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Synonym 5-methoxy-2-methylbenzothiazole, 5-methoxy-2-methylbenzo d thiazole, benzothiazole, 5-methoxy-2-methyl, 2-methyl-5-methoxybenzothiazole, 2-methyl-5-methoxy-benzothiazole, acmc-209h8o, cambridge id 5150005, benzothiazole,5-methoxy-2-methyl, 2-methyl-5-methoxy-1,3-benzothiazole, 2-methyl-5-methoxy-1, 3-benzothiazole
Formula Weight 179.24
Percent Purity ≥98.0% (GC)
Physical Form Lumps
Chemical Name or Material 5-Methoxy-2-methylbenzothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.