Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-methylindole, 99+%

Catalog No. AC125940050
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125940050 5 g Glass bottle
AC125940010 1 g Glass bottle
AC125940250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125940050 Supplier Thermo Scientific Chemicals Supplier No. 125940050
Only null left
Add to Cart

CAS: 1076-74-0 | C10H11NO | 161.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076-74-0
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005620
InChI Key VSWGLJOQFUMFOQ-UHFFFAOYSA-N
Synonym 5-methoxy-2-methylindole, 2-methyl-5-methoxyindole, 1h-indole, 5-methoxy-2-methyl, indole, 5-methoxy-2-methyl, pubchem7236, acmc-1buiu, maybridge1_002343, 5-methoxy-2 methylindole, 2-methyl-5-methoxylindole, 2-methyl-5-methoxy indole
PubChem CID 70642
IUPAC Name 5-methoxy-2-methyl-1H-indole
SMILES COC1=CC=C2NC(C)=CC2=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 85°C to 88°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, II, 47
Formula Weight 161.2
Percent Purity 99+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Methoxy-2-methylindole, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-066-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.