Learn More
5-Methoxy-2-methylindole, 99+%
Description
It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed as a reactant in preparation of indolylquinoxalines by condensation reactions, reactant in preparation of alkylindoles via Ir-catalyzed reductive alkylation, reactant in arylation reactions using a palladium acetate catalyst, reactant in enantioselective Friedel-Crafts alkylation and reactant in stereoselective synthesis of cyclopentaindolones via stereoselective [3+2] cyclopentannulation. As an indole derivative shown to be an effective inhibitor of the chlorinating activity of myeloperoxidase (MPO). Used in the metabolic synthesis of arylacetic acid anti-inflammatory synthesis.
Solubility
Soluble in methanol (very faint turbidity.)
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 85°C to 88°C |
| Quantity | 5 g |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in methanol (very faint turbidity.) |
| Formula Weight | 161.2 |
| Percent Purity | ≥99% |
| Chemical Name or Material | 5-Methoxy-2-methylindole |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 214-066-5 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.