Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1,3,4-thiadiazole-2-thiol 98.0+%, TCI America™
SDP

Catalog No. M0976250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0976250G 250 g
M097625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0976250G Supplier TCI America Supplier No. M0976250G

Chemical Identifiers

CAS 29490-19-5
Molecular Formula C3H4N2S2
Molecular Weight (g/mol) 132.20
MDL Number MFCD00038349
InChI Key FPVUWZFFEGYCGB-UHFFFAOYSA-N
Synonym 2-mercapto-5-methyl-1,3,4-thiadiazole, 5-methyl-1,3,4-thiadiazole-2-thiol, 1,3,4-thiadiazole-2 3h-thione, 5-methyl, 5-methyl-1,3,4-thiadiazole-2 3h-thione, 5-methyl-1,3,4-thiadiazol-2-thiol, 2-methyl-5-mercapto-1,3,4-thiadiazole, d2-1,3,4-thiadiazoline-5-thione, 2-methyl, cefazolin impurity e, pubchem9932, acmc-1cbwn
PubChem CID 1810203
IUPAC Name 5-methyl-2,3-dihydro-1,3,4-thiadiazole-2-thione
SMILES CC1=NNC(=S)S1

Specifications

Melting Point 186°C
Color White-Yellow
Quantity 250 g
Formula Weight 132.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-1,3,4-thiadiazole-2-thiol
EINECSNumber (5)-0569
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.