Learn More
Sigma Organic Chemistry 5-Methyl-1H-pyrazole | 50mg | MFCD08685900
Supplier: Sigma Organic Chemistry CDS00243050MG

5-Methyl-1H-pyrazole , 50mg
About this Item:
MDL #: MFCD08685900
Molecular Weight: 82.1
UNSPSC Code: 12352200
Chemical Formula: C4H6N2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.