Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile 97.0+%, TCI America™
SDP

Catalog No. M21385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21385G 5 g
M213825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21385G Supplier TCI America Supplier No. M21385G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 138564-59-7
Molecular Formula C12H9N3O2S
Molecular Weight (g/mol) 259.283
MDL Number MFCD06408054
InChI Key NPXUFPFFHANGDL-UHFFFAOYSA-N
Synonym 3-Cyano-5-methyl-2-(2-nitroanilino)thiophene
PubChem CID 395460
IUPAC Name 5-methyl-2-(2-nitroanilino)thiophene-3-carbonitrile
SMILES CC1=CC(=C(S1)NC2=CC=CC=C2[N+](=O)[O-])C#N

Specifications

Melting Point 111°C
Quantity 5 g
UN Number 3077
Formula Weight 259.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-(2-nitroanilino)-3-thiophenecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.