Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. M29351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29351G 1 g
M29355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29351G Supplier TCI America Supplier No. M29351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 956317-36-5
Molecular Formula C10H9N3O2
Molecular Weight (g/mol) 203.201
MDL Number MFCD18382679
InChI Key SRBAGFIYKNQXDV-UHFFFAOYSA-N
PubChem CID 53260165
IUPAC Name 5-methyl-2-(triazol-2-yl)benzoic acid
SMILES CC1=CC(=C(C=C1)N2N=CC=N2)C(=O)O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 1 g
Formula Weight 203.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-2-(2H-1,2,3-triazol-2-yl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.