Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-hexanone, 99%

Catalog No. AAL175840F
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL175840F 2500 mL
AAL17584AE 100 mL
AAL17584AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL175840F Supplier Thermo Scientific Chemicals Supplier No. L175840F
Add to Cart

CAS: 110-12-3 | C7H14O | 114.188 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. (5.4 g/L)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-12-3
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00008950
InChI Key FFWSICBKRCICMR-UHFFFAOYSA-N
Synonym 5-methyl-2-hexanone, 2-hexanone, 5-methyl, isoamyl methyl ketone, methyl isoamyl ketone, miak, isopentyl methyl ketone, 2-methyl-5-hexanone, ketone, methyl isoamyl, methyl isopentyl ketone, unii-6o4a4a5f28
PubChem CID 8034
IUPAC Name 5-methylhexan-2-one
SMILES CC(C)CCC(=O)C

Specifications

Melting Point -74°C
Density 0.812
Boiling Point 144°C to 145°C
Flash Point 41°C (105°F)
Refractive Index 1.4075
Quantity 2500 mL
UN Number UN2302
Beilstein 506163
Solubility Information Soluble in water. (5.4g/L)
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material 5-Methyl-2-hexanone
Show More Show Less
Hazard Category H226-H332
Hazard Statement GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 5-METHYLHEXAN-2-ONE
EINECSNumber 203-737-8
RTECSNumber MP3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.