Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate, 97%, Thermo Scientific™

Catalog No. CC00706CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC00706CB 250 mg
CC00706DA 1 g
CC00706DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC00706CB Supplier Thermo Scientific Chemicals Supplier No. CC00706CB
Add to Cart

Chemical Identifiers

CAS 306935-03-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
InChI Key WTKWFNIIIXNTDO-UHFFFAOYSA-N
Synonym 5-methyl-2-trifluoromethyl-3-furyl isocyanate, 3-isocyanato-5-methyl-2-trifluoromethyl furan, furan,3-isocyanato-5-methyl-2-trifluoromethyl, 5-methyl-2-trifluoromethyl fur-3-yl isocyanate, 3-isocyanato-5-methyl-2-trifluoromethyl-furan, 5-methyl-2-trifluoromethyl-3-furylisocyanate, 5-methyl-2-trifluoromethyl furan-3-isocyanate, furan, 3-isocyanato-5-methyl-2-trifluoromethyl-9ci
PubChem CID 2779896
IUPAC Name 3-isocyanato-5-methyl-2-(trifluoromethyl)furan
SMILES CC1=CC(=C(O1)C(F)(F)F)N=C=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 191.11
Percent Purity 90%
Chemical Name or Material 5-Methyl-2-(trifluoromethyl)-3-furyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.