Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-4-isoxazolecarbonyl chloride, 97%, Thermo Scientific™

Catalog No. CC26302DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC26302DA 1 g Amber Glass Bottle
CC26302CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC26302DA Supplier Thermo Scientific Chemicals Supplier No. CC26302DA

Chemical Identifiers

CAS 67305-24-2
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.542
InChI Key ZKAQPVQEYCFRTK-UHFFFAOYSA-N
Synonym 5-methyl-4-isoxazolecarbonyl chloride, 5-methylisoxazole-4-carbonyl chloride, 5-methyl-4-isoxazolecarbonylchloride, 4-isoxazolecarbonyl chloride, 5-methyl, 4-isoxazolecarbonylchloride, 5-methyl, 5-methyl-isoxazole-4-carbonyl chloride, acmc-209nyg, methyl-4-isoxazolecarbonyl chloride, 4-isoxazolecarbonylchloride,5-methyl
PubChem CID 2759917
IUPAC Name 5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES CC1=C(C=NO1)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.54
Percent Purity 97%
Chemical Name or Material 5-Methyl-4-isoxazolecarbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.