Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl L-Glutamate 98.0+%, TCI America™
SDP

Catalog No. M169025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M169025G 25 g
M16905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M169025G Supplier TCI America Supplier No. M169025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1499-55-4
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.157
InChI Key ZGEYCCHDTIDZAE-BYPYZUCNSA-N
Synonym l-glutamic acid 5-methyl ester, h-glu ome-oh, polymethylglutamate, 2s-2-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid gamma-methyl ester, 5-methyl l-glutamate, l-glutamic acid, 5-methyl ester, 2s-2-amino-5-methoxy-5-oxo-pentanoic acid, 2s-2-amino-4-methoxycarbonyl butanoic acid
PubChem CID 68662
IUPAC Name (2S)-2-amino-5-methoxy-5-oxopentanoic acid
SMILES COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 25 g
Formula Weight 161.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl L-Glutamate
EINECSNumber (2)-1312&(9)-0418
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.