Learn More
AdipoGen 5-Methylmellein
SDP
Supplier:  AdipoGen BVT0413M001
Encompass_Preferred
Chemical. CAS 7734-92-1. Formula C11H12O3. MW 192.2. Isolated from Fusicoccum amygdalis. Widespread fungal pentaketide. Weak antibacterial, antifungal and antiviral activity. Specific inhibitor of human DNA polymerase lambda. Weak antigerminative activity.
Catalog No. 50-256-1942
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.