Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™

Catalog No. AC158672500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC158672500 250 mg
AC158670010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158672500 Supplier Thermo Scientific Chemicals Supplier No. 158672500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C11H15N2
Molecular Weight (g/mol) 175.25
MDL Number MFCD00012683
InChI Key PYOUAIQXJALPKW-UHFFFAOYSA-O
Synonym 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287
PubChem CID 6917145
SMILES CC1=CC=C2NC=C(CC[NH3+])C2=C1

Specifications

CAS 55795-89-6
CAS Min % 97.5
CAS Max % 100.0
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 250 mg
IUPAC Name 2-(5-methyl-1H-indol-3-yl)ethan-1-aminium
Formula Weight 210.71
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyltryptamine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.