Learn More
Eisco™ 5 mL Glass Volumetric Flask with Blue Graduations and Stopper
Catalog No. 12183036 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric): 5 mL
Catalog No.
12-183-036 Capacity (Metric): 5 mL
1 options
Catalog No. 12-183-036 Supplier Eisco™ Supplier No. CH1972AST
Volumetric Flask, 5 mL, Class A, ASTM. With a polypropylene stopper and a blue graduation. Made of Borosilicate Glass
  • Includes individual work certificate
  • Class A tolerance
Specifications
Capacity (Metric) 5 mL
Material Borosilicate Glass (3.3)
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated No
Mouth Style Beaded
Sterility Non-sterile
Class Class A
Bottom Shape Flat
Surface Treatment Uncoated