Learn More
Description
- Matched to Bruker™ turbines
- Tachometer strip improves life span
- Temperature range: -20 to 60 °C
Specifications
Specifications
| For Use With (Equipment) | Bruker NMR Spectrometer |
| Type | Spinner Turbine |
| Color | Black |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.